Proto® Heat Shrink Loops, Stanley® Products
Supplier: Safety & Industrial Supplies
577-P2R275X2LP
76191-174EA 198.35 USD
76191-174
Proto® Heat Shrink Loops, Stanley® Products
Fall Protection Fall Protection Kits
Easy installation, quick attachment to lanyards, and elimination of confusion with matching D-ring webbing to proper shrink rings.
  • For use with tool lanyards and tethers
  • Installation via heat gun
  • Superior strength over conventional heat shrink methods
Designed to keep the stitching away from the heat source to prevent damage to the stitching common with conventional methods. Prevents webbing from slipping out of the heat shrink. Allows for proper labeling of each shrink loop containing load limits, usage instructions, and precautions.