Proto® Torqueplus™ Metric Sockets, ³/₈", ORS Nasco
Supplier: Safety & Industrial Supplies
577-52055MH
76190-768EA 9.57 USD
76190-768
Proto® Torqueplus™ Metric Sockets, ³/₈", ORS Nasco
Wrenches
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners.
  • Metric
  • Forged alloy steel
  • Ball locking
  • Square drive
  • Nickel chrome finish
Proto Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.