Starting Punch5 5/8 in 3/16 in tip
Supplier: ORS NASCO, INC.
Starting Punch, type: Tapered, Material: Tool Steel, Overall Length: 5 5/8 in, Stock Shape: Hex, Tip Size: 3/16 in, Tip Type: Round, Heavier, shorter body and wider point than drift punch, For installing pins and rivets