Proto® Tether-Ready Drive Deep Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-5006M-TT 577-5007M-TT 577-5008M-TT 577-5008-TT 577-5009M-TT 577-5019M-TT 577-5020M-TT 577-5021M-TT 577-5022M-TT 577-5022-TT 577-5023M-TT 577-5024-TT 577-5026-TT 577-5217M-TT 577-5218M-TT 577-5218-TT 577-5219M-TT 577-5220M-TT 577-5220-TT 577-5221M-TT 577-5222M-TT 577-5222-TT 577-5223M-TT 577-5224M-TT 577-5010M-TT 577-5015M-TT 577-5016M-TT 577-5016-TT 577-5017M-TT 577-5018M-TT 577-5224-TT 577-5225M-TT 577-5226M-TT 577-5226-TT 577-5228-TT 577-5230-TT 577-5232-TT 577-5316-TT 577-5318-TT 577-5320-TT 577-5322-TT 577-5324-TT 577-5326-TT 577-5328-TT 577-5330-TT 577-5332-TT 577-5334-TT 577-5336-TT 577-5338-TT 577-5340-TT 577-5342-TT 577-5410M-TT 577-5411M-TT 577-5412M-TT 577-5412-TT 577-5413M-TT 577-5414M-TT 577-5414-TT 577-5415M-TT 577-5416M-TT 577-5416-TT 577-5417M-TT 577-5418M-TT 577-5418-TT 577-5419M-TT 577-5420M-TT 577-5420-TT 577-5421M-TT 577-5422M-TT 577-5422-TT 577-5423M-TT 577-5426-TT 577-5427M-TT 577-5428-TT 577-5429M-TT 577-5430M-TT 577-5430-TT 577-5431M-TT 577-5432-TT 577-5434-TT 577-5436-TT 577-5438-TT 577-5440-TT 577-5442-TT 577-5444-TT 577-5010-TT 577-5011M-TT 577-5012M-TT 577-5013M-TT 577-5014M-TT 577-5206M-TT 577-5207M-TT 577-5208M-TT 577-5208-TT 577-5209M-TT 577-5210M-TT 577-5210-TT 577-5211M-TT 577-5212M-TT 577-5212-TT 577-5213M-TT 577-5214M-TT 577-5214-TT 577-5215M-TT 577-5216M-TT 577-5216-TT 577-5312-TT 577-5314-TT 577-5424M-TT 577-5424-TT
76189-128EA 44.37 USD
76189-128 76189-130 76189-132 76189-134 76189-136 76189-144 76189-148 76189-152 76189-154 76189-156 76189-158 76189-160 76189-164 76189-234 76189-236 76189-238 76189-240 76189-244 76189-246 76189-250 76189-252 76189-254 76189-258 76189-260 76189-414 76189-418 76189-420 76189-422 76189-424 76189-428 76189-430 76189-434 76189-436 76189-438 76189-440 76189-442 76189-444 76189-464 76189-466 76189-470 76189-472 76189-476 76189-478 76189-480 76189-482 76189-484 76189-486 76189-488 76189-490 76189-492 76189-494 76189-526 76189-530 76189-534 76189-536 76189-538 76189-540 76189-542 76189-544 76189-546 76189-548 76189-552 76189-554 76189-556 76189-558 76189-560 76189-564 76189-570 76189-572 76189-574 76189-576 76189-580 76189-582 76189-586 76189-588 76189-590 76189-592 76189-594 76189-598 76189-602 76189-606 76189-608 76189-610 76189-652 76189-654 76190-734 76190-736 76190-740 76190-744 76190-748 76190-770 76190-772 76190-774 76190-776 76190-778 76190-780 76190-782 76190-786 76190-790 76190-792 76190-794 76190-796 76190-798 76190-800 76190-802 76190-804 76190-850 76190-852 76190-854 76190-856
Proto® Tether-Ready Drive Deep Sockets, ORS Nasco
The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
  • Manufactured with four detent holes; pin detent will engage correctly regardless of the direction the socket is placed on the square drive
  • Positive Control System requires a separate tool to attach or remove the socket, meaning it cannot be removed without specific intent
  • Chrome finish is applied after the holes are bored for better corrosion resistance
  • Ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials
The TorquePlus™ configuration employs a radius-corner design, shifting contact area and stress away from the fastener corners and distributing contact stresses over a large area of the socket, minimizing slippage and fastener rounding, helping to extend fastener life, and allowing the user to more easily turn worn, rounded, or rusted fasteners.