Proto® Center Punches, ORS Nasco
Supplier: Safety & Industrial Supplies
577-413/8 577-415/16 577-415/8 577-417/16 577-411/2 577-411/4
76188-816EA 13.18 USD
76188-816 76188-818 76188-820 76188-822 76190-688 76190-690
Proto® Center Punches, ORS Nasco
Chisels
Modified parabolic striking end for greater resistance to mushrooming and chipping.
  • Center punch
  • Round tip
  • Hex stock shape
  • Tool steel
For metal working layout such as marking intersecting lines, hole locations, and pilot drill starting points.