Proto® Leverage Wrenches, Essendant
Supplier: Safety & Industrial Supplies
577-2632PW 577-2617PW
76188-668EA 194.75 USD
76188-668 76188-712 76190-636
Proto® Leverage Wrenches, Essendant
Wrenches
Leverage wrenches provide a safe alternative to striking wrenches in confined spaces and on applications that prohibit sparking.
  • Offset box wrench head offers access to recessed fasteners and keeps hands clear of obstructions
  • Ergonomic design allows wrenches to be used conventionally without the handle
  • The handle can be attached to provide increased leverage for final tightening
  • Bar stop on the wrench shaft helps to align locking button with handle hole
  • Ideal for oil and gas industries