Proto® Rolling Head Pry Bar Set, ORS Nasco
Supplier: Safety & Industrial Supplies
577-2138
76190-562EA 236.03 USD
76190-562
Proto® Rolling Head Pry Bar Set, ORS Nasco
Crow Bars
Pry and aligning bar with a pointed taper and 90° rolling head for leverage.
  • High alloy steel is heat treated the entire length, and black oxide treated for added strength and durability