Proto® Torqueplus™ 12-Point Combination Wrenches, Satin Finish, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1211A
76190-162EA 26.38 USD
76190-162
Proto® Torqueplus™ 12-Point Combination Wrenches, Satin Finish, ORS Nasco
Wrenches
Torqueplus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Oval handle
  • Open/box opening type
  • Inch measuring system
  • Forged alloy steel
  • Satin finish
Reduces risk of fastener rounding. Applies more torque to fastener. Unique arc design increases surface contact. Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury. Shank is positioned at the center of box end, reduces the tendency of the wrench to cock.