Proto® Torqueplus™ Impact Sockets, ¹/₂", ORS Nasco
Supplier: Safety & Industrial Supplies
577-7450H
76189-814EA 52.81 USD
76189-814
Proto® Torqueplus™ Impact Sockets, ¹/₂", ORS Nasco
Wrenches
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners
  • Forged alloy steel
  • Square drive
  • Black oxide finish
  • Pin locking
  • Inch measuring
Proto Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.