Proto® Brass Hammer, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1432G-TT
76189-316EA 195.79 USD
76189-316
Proto® Brass Hammer, ORS Nasco
Mallets and Hammers
Mar-resistant soft brass head helps absorb impact and reduces work damage.
  • Spark-resistant for safe use near combustible materials
  • Fiberglass handle with ergonomic, non-slip grip