Proto® Torqueplus™ Metric Sockets, ¹/₄", Stanley® Products
Supplier: Safety & Industrial Supplies
577-4704TM
76188-982EA 10.76 USD
76188-982
Proto® Torqueplus™ Metric Sockets, ¹/₄", Stanley® Products
Wrenches
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
  • Forged alloy steel
  • Metric
  • Ball locking type
  • Square drive
  • Nickel chrome finish
Proto Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.