Proto® Long Aligning Punches, ORS Nasco
Supplier: Safety & Industrial Supplies
577-155/8
76188-638EA 38.41 USD
76188-638
Proto® Long Aligning Punches, ORS Nasco
Chisels
Modified parabolic striking end for greater resistance to mushrooming and chipping.
  • Tool steel
  • Tapered punch
  • Round tip
  • Hex stock shape
For lining up holes in flanges, assemblies, and for other mating parts and for removing long pins and shafts.