Wolverine Thin Cutting Wheels, Weiler
Supplier: Safety & Industrial Supplies
804-56281
76184-170EA 2.84 USD
76184-170
Wolverine Thin Cutting Wheels, Weiler
Abrasives
Resin-bonded disc is formulated for aggressive stock removal and fast cutting
  • Reinforced with fiberglass webbing for stability
  • Use with a thin cutting wheel, cut-off die grinder, or right angle air die grinder
  • Use for cutting and parting, notching, rough grinding, and weld removal applications
Suitable for general purpose cutting and grinding applications on steel, iron, and other ferrous metals.