Knot Wire Bevel Wheel, Weiler®
Supplier: Safety & Industrial Supplies
804-13456
76183-788EA 47.19 USD
76183-788
Knot Wire Bevel Wheel, Weiler®
Abrasives
Maximum aggression for cleaning in corners.
  • Heavy-duty brushing action with some flexibility
  • Ideal for cleaning filet welds
  • Steel filter material
Use with right-angled grinders to remove rust, paint, oxides, and scale.