Wide Face Standard Twist Knot Wire Wheel, Weiler
Supplier: Safety & Industrial Supplies
804-09180
76183-768EA 160.84 USD
76183-768
Wide Face Standard Twist Knot Wire Wheel, Weiler
Abrasives
Rugged knot-type construction provides aggressive, high-impact action for severe applications.
  • Heavy-duty brushing action with some flexibility
  • Double row of knotted wire for a wider brush face
  • Use with bench/pedastal grinders
  • Steel filter material
Use for coating removal, heavy deburring, removal of excess molded rubber, heavy corrosion, rust, paint, oxides, and scale removal, and surface roughening and preparation applications.