Wide Face Crimped Wire Wheel, Weiler
Supplier: Safety & Industrial Supplies
804-03210
76183-714EA 311.73 USD
76183-714
Wide Face Crimped Wire Wheel, Weiler
Abrasives
Maximum wire density and aggression, covers more surface area in a single pass
  • Provides flexible brushing action
  • Use with bench/pedastal grinders
  • Steel filter material
Removes rust, paint, oxides, and scale.
Consistent and effective performance for cleaning, light deburring, edge blending, roughening, and finishing.