Automatic Feeder, LV12
Supplier: Mettler Toledo
30374389
76183-634EA 15249.41 USD
76183-634
Automatic Feeder, LV12
Sample Dividers
Reliable item separation.
  • Automate weighing process for tablets
  • Speed up statistical quality control
  • Remote controlled by balance
  • Adjustable speed
Feeds tablets and small parts unattended and reliable piece by piece into the weighing pan. Shortens the time required for weighing large samples of tablets. Process controlled by connected SQC balance or device.
The control interface to the balance ensures synchronized feeding of items to the balance.