CytoFLEX Sheath Fluid, Beckman Coulter
Supplier: BECKMAN COULTER
B51503
76183-428EA 41.24 USD
76183-428
CytoFLEX Sheath Fluid, Beckman Coulter
Cell Culture Supplements and Additives
CytoFLEX Sheath Fluid is a nonionic, non-fluorescent, and azide-free sheath fluid for use on Beckman Coulter CytoFLEX flow cytometer.
The diluent is composed of high quality water with an antimicrobial compound.