Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer
Supplier: Antyila Scientific
36225-91
76183-396EA 444.81 USD
76183-396
Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer
Controllers Temperature Controllers
Manually set power control dial from 5 to 100% to control the output wattage.
  • Simplify temperature control with repeating timed On/Off cycles
  • Pilot light on front lets users know when the load is heating
Ordering information: Includes 6 ft. attached power cord.