Digi-Sense® Benchtop Variable Voltage Output Controller, Cole-Parmer
Supplier: Antyila Scientific
36225-59
76183-394EA 495.52 USD
76183-394
Digi-Sense® Benchtop Variable Voltage Output Controller, Cole-Parmer
Controllers Utility Controllers
Designed to regulate the input voltage through the selector knob.
  • Works with resistive loads, providing variable voltage
  • Simple to use; no programming or feedback required
  • With reference knob (0 to 100%)
Output 1: Proportional Voltage
Ordering information: Includes 6 ft. attached power cord.