Digi-Sense® Pre-Calibrated Humidity and Temperature Indicator, Cole-Parmer
Supplier: Cole-Parmer Instrument Co.
2025030
76183-376EA 51.18 USD
76183-376
Digi-Sense® Pre-Calibrated Humidity and Temperature Indicator, Cole-Parmer
Hygrometers Digital Thermo-Hygrometers
Designed to gauge proper humidity levels.
  • Reliable measurements guaranteed
  • Pre-calibrated to NIST-traceable standards
  • Unique environment comfort indicator
  • Displays both temperature and humidly
Model 20250-30 has built-in temperature and humidity sensors designed for indoor measurements.
Humidity accuracy: ±5% RH; ±8% RH above 80% and below 40% RH
Certifications: NIST-traceable calibration certificate.
Delivery information: Includes built-in desktop stand, wall-mounting bracket, one AAA battery, and NIST-traceable calibration report with data supplied by InnoCal.