Digi-Sense® Pre-Calibrated Thermo Hygrometers, Cole-Parmer
Supplier: Antyila Scientific
20250-11
76183-362EA 215.94 USD
76183-362
Digi-Sense® Pre-Calibrated Thermo Hygrometers, Cole-Parmer
Hygrometers Digital Thermo-Hygrometers
Ideal for use in the HVAC, IAQ, and environmental industries.
  • Reliable measurements guaranteed
  • Pre-Calibrated to NIST-traceable standards
  • ±2% basic RH accuracy
Model 2025011 features a backlit LCD, wet bulb temperature, and absolute humidity. The humidity accuracy for this model is ±2% RH from 40% to 60% RH; ±3% RH from 20% to 40% RH and 60% to 80% RH; ±4% RH from 0% to 20% RH and 80% to 100% RH. Model 2025012 features one channel, mini ANSI connector, backlit LCD, dry bulb and wet bulb temperature, and TSH/TEET functions. The humidty accuracy for this model is ±2.5% RH; ±5% above 90% and below 10% RH.
Certifications: NIST-traceable calibration certificate with data, performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.
Delivery information: Includes carrying case, three AAA batteries, and NIST-traceable calibration report with data supplied by InnoCal. Model 20250-12 also includes a Type-K flexible probe.