Digi-Sense® Pre-Calibrated Professional Infrared Thermometers, Cole-Parmer
Supplier: Cole-Parmer Instrument Co.
2025007 20250-08
76183-358EA 468.75 USD
76183-358 76183-360
Digi-Sense® Pre-Calibrated Professional Infrared Thermometers, Cole-Parmer
Thermometers Digital Thermometers
Measure surface temperatures quickly and accurately. Safely and easily locate hot and cold spots, preventing problems and saving costs.
  • Reliable measurements guaranteed
  • Pre calibrated to NIST-traceable standards
  • High distance-to-sight (D/S) ratios handle long-distance measurements
  • External Type-K input for contact measurements
  • Dual-laser sighting to pinpoint target areas
  • Protective lens cover with magnetic lock
  • Pistol-grip handle is comfortable and easy to use
  • USB interface for easy data transmission
  • Adjustable emissivity, backlit LCD
Temperature accuracy for the 30:1 D/S model is ±5.4 °F from -58° to 68 °F; ±1.0% of reading ± 1.8 °F from 68° to 752 °F; ±1.5% of reading ± 3.6 °F from 752° to 1472 °F; ±2.5% of reading from 1472° to 3002 °F (±3.0 °C from -50° to 20 °C; ±1.0% of reading ± 1.0 °C from 20° to 400 °C; ±1.5% of reading ± 2.0 °C from 400° to 800 °C; ±2.5% of reading from 800° to 1650 °C). Temperature accuracy for the 50;1 D/S model is ±5.4 °F from -58° to 68 °F; ±1.0% of reading ± 1.8 °F from 68° to 932 °F; ±1.5% of reading ± 3.6 °F from 932° to 1832 °F; ±2.0% of reading from 1832° to 3362 °F (±3.0 °C from -50° to 20 °C; ±1.0% of reading ± 1.0 °C from 20° to 500 °C; ±1.5% of reading ± 2.0 °C from 500° to 1000 °C; ±2.0% of reading from 1000° to 1850 °C).
Certifications: NIST-traceable calibration certificate with data, performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.
Delivery information: Includes Type-K flexible probe, USB cable, software, tripod, carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.