Supplier: SPEX
2600
76181-832EA 4766.74 USD
76181-832
Homogenizers Tissue Grinders
Hold temperature sensitive samples chilled prior to grinding.
  • Fill insulated jacket manually or automatically with liquid nitrogen supply
  • Quickly reach liquid nitrogen temperatures
Cryo-Station chamber offers a convenient place to keep samples that must be kept chilled prior to grinding or to preserve pesticides, RNA, or proteins for extraction. The well can hold a chilled cutting board (included) to prepare temperature sensitive samples.