Cole-Parmer® HG-400 MiniG® Tissue Homogenizer and Cell Lyser
Supplier: SPEX
1600
76181-800EA 15111.95 USD
76181-800
Cole-Parmer® HG-400 MiniG® Tissue Homogenizer and Cell Lyser
Homogenizers Benchtop Homogenizers
The Cole-Parmer® HG-400 MiniG® is small yet powerful tissue homogenizer and cell lyser.
  • Ideal for cell lysis applications including DNA/RNA extraction, protein extraction, and pesticide residue analysis
  • The grinding process is visible through the see-through lid window
  • Comes with an adjustable clamp that allows users to secure vials or microplates of various sizes
  • Operation is quick and easy
  • Digital display allows for control of the rate and time
The adjustable clamp can handle sample vials from 2 to 50 ml or up to two deep-well microplates. Using the bead beating method, it is designed for rapid cell disruption, cell lysis, and tissue homogenization. Common applications for this model include DNA/RNA research and extraction, protein extraction, biofuel research, and pesticide residue extraction via the QuEChERS or SWeET method.
Certifications: CE approved.