Heavy-Duty Flexible Drum Heaters with Thermostat, Cole-Parmer
Supplier: Antyila Scientific
03106-65 03106-66 03106-67 03106-68 03106-73 03106-74 03106-75 03106-76
76181-542EA 230.86 USD
76181-542 76181-682 76181-684 76181-686 76181-688 76181-690 76181-692 76181-694
Heavy-Duty Flexible Drum Heaters with Thermostat, Cole-Parmer
Heaters Heating Mantles
Wrap around a metal or plastic drum to maintain temperature and viscosity.
  • Extra-wide 4" heaters provide better surface coverage
  • Resistant to moisture and chemicals
  • US Standard plug type
  • Minimum temperature 50 °F
The built-in thermostat provides adjustable temperature control.