BriskHeat® 55-Gallon Drum Cover, Antylia Scientific
Supplier: Antyila Scientific
09300-70
76181-588EA 177.5 USD
76181-588
BriskHeat® 55-Gallon Drum Cover, Antylia Scientific
Covers
Prevent heat loss from your drum heater.
  • Suitable for use with the Briskheat® 55-gallon insulated drum heaters
This BriskHeat 55-gallon drum insulation top cover is used to minimize heat loss and speed up heat-up time. Made to fit standard 55-gallon drums with cut-outs to access plug and bung without removing cover.