MicroSampler, Burkle
Supplier: BURKLE INC
5307-0102 5307-0105 5307-0110 5307-0202 5307-0205 5307-0210 5307-1120 5307-2055 5307-2085 5307-2120 5307-1055 5307-1085
76181-222EA 165.11 USD
76181-222 76181-224 76181-226 76181-228 76181-230 76181-232 76181-250 76181-252 76181-254 76181-256 76181-436 76181-438
MicroSampler, Burkle
Samplers Solid Samplers
The MicroSampler has been specially designed for the sampling of small quantities of powder.
  • Point sampling
  • Various lengths and volumes are available
  • Tip can be removed for easy cleaning
  • Stainless steel AISI 316L (1.4404)
  • Sterilizable
  • Additional tips are available in different sizes
Volumes of 0.2 mL (0.01 oz.), 0.5 mL (0.02 oz.), 1.0 mL (0.03 oz.), 2.0 mL (0.07 oz.), 5.0 mL (0.17 oz.), and 10 mL (0.34 oz.) can be sampled easily, quickly, and cleanly at different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable, allowing tips with different volumes to be used.
To use, simply insert the MicroSampler into powder. At the predetermined point, open the volume tip by pressing the button, allowing the sample to flow in. Close the volume tip once the sample is collected. Pull back the lance, open the volume tip again, and empty the sample for further measurement or analysis.
Ordering information: Lance and head should be ordered separately.