TBE 10X Buffer, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

11TBE10X02
76180-758EA 93.42 USD
76180-758
TBE 10X Buffer, MP Biomedicals
Buffers Electrophoresis Buffers Running Buffers

TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis, after dilution to working concentrations.


  • Use with non-denaturing or denaturing gels
  • pH: 8.2 - 8.4 (Lit.)
  • Soluble in water


TBE buffer maintains the structural integrity of nucleic acids and more suitable for their size analysis. TBE has a greater buffering capacity and will give sharper resolution. TBE is better suited for high-voltage (>150 V) electrophoresis because of its higher buffering capacity.


TBE is routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a31c54d3c0d0c0a991e6c5c7fe530ad1c50ffa36&vid=d8458fe6df1aedc3e58fbb731f8b580915fbeb2a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR