TAE 50X Buffer, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11TAE50X01
76180-756EA 163.43 USD
76180-756
TAE 50X Buffer, MP Biomedicals
Buffers Electrophoresis Buffers Running Buffers
Buffer for Electrophoresis.
  • Used to prepare buffers for polyacrilamide and Agarose gel electrophoresis.
  • Components: Tris Base 2M; Acetic acid 1M; EDTA 0,05 M
  • Filtered through 0.2pm PTFE aseptically filled
  • Conductivity of 1X conc. (25°C): 1.3 mS/cm Conductivity of 50X conc. (25°C): 18 mS/cm
TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abea99142d973b991ba08568ec73658e9184973d&vid=eff55f45a5314cb34632b9a4e7476e0eb8902224&prid=web&clid=SAPDAM