SSC 20X Buffer, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11RIST2050
76180-744EA 83.98 USD
76180-744
SSC 20X Buffer, MP Biomedicals
Buffers Electrophoresis Buffers Hybridization Buffers
SSC Buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates.
  • Molecular Biology Grade
  • Clear, Colorless Solution
  • 20x Concentrated
  • Formulation: 3 M Trisodium citrate
  • Sterility: 0.2 μm filtered
SSC is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.