Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Used to minimize cell attachment or aggregation.
- Sterile
- Suitable for Cell Culture
- Formulation: Inorganic Salts: Potassium Chloride [KCl] 400.00000 mg/L, Potassium Phosphate Monobasic [KH2PO4] 60.00000 mg/L, Sodium Bicarbonate [NaHCO3] 350.00000 mg/L, Sodium Chloride [NaCl] 8000.00000 mg/L, Sodium Phosphate Dibasic [Na2HPO4] 47.50000 mg/L, Dextrose 1000.00000 mg/L, Phenol Red Sodium Salt 17.00000 mg/L
- pH: 6.9-7.4
- Osmolality 258 - 298 mM/kg
Calcium and magnesium support cell adhesion. Hanks' Balanced Salts (Modified), without Magnesium and Calcium, is used to minimize cell attachment or aggregation.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...