CARROUSEL™ Pipette Stand, Gilson
Supplier: GILSON, INC.
F161401
76178-564EA 170.13 USD
76178-564
CARROUSEL™ Pipette Stand, Gilson
Pipette Stands
The CARROUSEL™ Pipette Stand holds up to seven Gilson single or multichannel pipettes, protecting them from contamination.
  • Pipettes hang vertically from the rack, preventing liquids from entering and contaminating the pipette shaft
  • Made from chemically resistant polypropylene and polycarbonate
  • Stand’s heavy base provides added stability
The rotating head of the CARROUSEL Stand provides quick access to pipettes while keeping your benchtop decluttered. Its space-saving design frees your bench space for more important tasks at hand.