DISTRIMAN® Repetitive Pipettor, Gilson®
Supplier: GILSON, INC.
F164001
76178-538EA 867.67 USD
76178-538
DISTRIMAN® Repetitive Pipettor, Gilson®
Pipettes Repetitive Pipettors
Adjustable repetetive positive-displacement pipette DISTRIMAN®.
DISTRIMAN® is a positive-displacement, continuously adjustable, repetitive pipette with direct read out designed to simplify multiple dispensing when used with DISTRITIPS® syringes. With DISTRIMAN, you can dispense the exact volume required for any protocol, even fractional volumes. It is ideal for repetitive dispensing in clinical, hospital, biological, chemical, food and beverage, pharmaceutical, cosmetics, and forensics labs.