Polysorbate 20, clear cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02194724.5
76177-882EA 82.27 USD
76177-882
Polysorbate 20, clear cell culture reagent
Polysorbate 20

Storage: Room Temperature
Polysorbate 20 is nonionic surfactants derived from sorbitan ester, it is soluble or dispersible in water but differ widely in organic and oil solubility.
Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharmaceutical applications. It is also used in pre-extraction of membranes to remove peripheral proteins & as a blocking agent on nitrocellulose at a typical concentration of 0.05%.
Series of nonionic surfactants derived from sorbitan esters. They are soluble or dispersible in water but differ widely in organic and oil solubilities. Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc.


  • Cell Culture Reagent Grade
  • Viscosity: 400cps
  • Refractive Index: n20/D 1.468 (lit.)
  • Presentation: Clear, Yellow to Amber Viscous Liquid
  • Critical Micelle Concentration (CMC): 0.06mM

Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ded0e706d3f5f5825efc6aa57017762c86e00470&vid=54bec7da01a35de0c9e7cdf43a931d5537510ba8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test A Passes
Identity Test B Passes
FTIR Conforms to Reference
Solubility (1g/20ml) Clear, Foamy Solution
Specific Gravity 1.08-1.13
Water Content ≤3%
Saponification Number 40-50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR