Dexamethasone

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

02199350.6
76177-524EA 140.76 USD
76177-524
Dexamethasone
Dexamethasone

An anti-inflammatory glucocorticoid with a range of effects on cell survival, cell signaling and gene expression. Use to study apoptosis, cell signaling pathways and gene expression.
Room Temperature


  • Water Soluble Grade
  • Presentation: White Powder
  • Greater than 55 mg dexamethasone per gram with a balance consisting of 2-hydroxypropyl-β-cyclodextrin. Typically used in cell culture applications.
  • Soluble in water (25 mg/ml - clear to slightly hazy, colorless solution)

Formula: C₂₂H₂₉FO₅
Storage Temperature: Ambient
MDL Number: MFCD00064136
CAS Number: 50-02-2

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR