Mineral oil light, liquid for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219483605 0219483625 0219483690
76177-358EA 25.79 USD
76177-358 76177-360 76177-362
Mineral oil light, liquid for molecular biology
Mineral oil light

Mineral Oil is a colorless, odorless mixture of alkanes from a mineral source. It is often used as an inert overlay to minimize evaporation in molecular biology applications, including PCR, but has many other applications on other research fields.


  • Refractive Index: n20/D 1.467 (lit.)


Mineral oil is suitable for use as an overlay to control evaporation and cross contamination of samples in a variety of molecular biology applications.

Boiling Pt: 218...643 °C (1013 hPa) (lit.)
Density: 0.81…0.894 (15 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR