Sodium dodecyl sulfate (SDS) 20% in aqueous solution , 99.5%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

0219895780 0219895790
76177-284EA 62.76 USD
76177-284 76177-286
Sodium dodecyl sulfate (SDS) 20% in aqueous solution , 99.5%
Sodium dodecyl sulphate (SDS)

SDS is an anionic detergent typically used to solubilize and denature proteins for electrophoresis.
SDS is used to solubilize and denature protein for Protein Electrophoresis & Blotting. SDS has also been used in large-scale phenol extraction of RNA to promote the dissociation of protein from nucleic acids when extracting from biological material. Surfactant used for mild, efficient, and greener dethioacetalization.
Store at Room Temperature (15-30 °C).


  • Concentration: 20%
  • Presentation: Liquid
  • Formulation: 20% SDS; 80% water
  • Aggregation Number: 62
  • Critical Micelle Concentration (CMC): 8.27mM
  • Soluble in water (200 mg/ml - clear, faint yellow solution), and ethanol (0.1g/10 ml)

Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1.03 g/mL (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d57a802560fb08de6d50e7f60e1cc21aab8a542b&vid=b02c4b8c7f7046254ff502cc279cf97d68440ef8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR