2,6,10,14-Tetramethylpentadecane

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Pristane

0219545505
76177-268EA 206.48 USD
76177-268
2,6,10,14-Tetramethylpentadecane
2,6,10,14-Tetramethylpentadecane

Pristane is a natural saturated terpenoid alkane obtained primarily from shark liver oil.
Pristane is used as a biological marker. It known to induce autoimmune diseases in rodents. It is used in research to understand the pathogenesis of rheumatoid arthritis and lupus, as a non-antigenic arthritogenic adjuvant and plasmacytomas.
Store at Room Temperature(15-30 °C)


  • Presentation: Clear, Colorless Liquid
  • Soluble in ether, petroleum ether, benzene, chloroform, carbon tetrachloride.

Formula: C₁₉H₄₀
MW: 268.53 g/mol
Boiling Pt: 296 (lit.)
Density: 0.783 g/mL (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008952
CAS Number: 1921-70-6
Merck Index: 12,07932

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR