Wizard™ CompactDry™ Reader, Hardy Diagnostics
Supplier: Hardy Diagnostics
CDR1
76177-196EA 13279.4 USD
76177-196
Wizard™ CompactDry™ Reader, Hardy Diagnostics
Colony Counters
Bring automation, efficiency and data security into your lab.
  • Automatically reads CompactDry™ plates in less than four seconds – stand alone system, no PC required
  • Counts and differentiates colonies based on the CompactDry™ type
  • Indexes cassette directly under the imaging camera for rapid reading
  • Accurately reads up to 150 colonies per plate
  • Accepts individual plates or groups of two, three, or four plates so you can keep your dilution series together - no need to seperate plates
  • Unit is suited for ambidextrous operation; plates can be scanned from the left or right side
  • Digitalized plate image can be saved to USB or exported to the user's LIMS program or included software management program
The Wizard™ CompactDry™ Reader is a fast, easy, and accurate way to read and record your CompactDry™ results. The Wizard™ has selectable modes in the configuration menu designed to read the different types of CompactDry™ plates.
Accessories information: Available accessories include USB wireless keyboard, USB barcode scanner, and USB thermal printer. Optional keyboard accessory for Wizard™ CompactDry™ Reader connects to Wizard™ by wireless USB dongle. Barcode reader for The Wizard™ CompactDry™ Reader connects via USB. Thermo Printer Option for the Wizard™ CompactDry™ Reader connects via USB.