CosMCPrep Plasmid Purification Kit, Beckman Coulter
Supplier: BECKMAN COULTER
A29174
76177-115EA 4927.1 USD
76177-115
CosMCPrep Plasmid Purification Kit, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
A SPRI paramagnetic bead-based plasmid purification system that uses a single protocol to purify a variety of high and low copy number template types. This flexible system is easily automated without sacrificing product quality.
  • Automated available
The single protocol, ease of automation and high quality results make CosMCPrep an ideal solution for all your plasmid purification needs. Kit components: L2 buffer, N3 buffer, pur4 buffer and RE1 buffer for 4000 preparations.