ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]), light green powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulfonate), ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

0219502301
IC19502305EA 242.56 USD
IC19502305 76177-058
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]), light green powder
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

2,2-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic acid) (ABTS) has been used as a chromogenic substrate for horseradish peroxidase (HRP), both in general activity assays and in ELISA applications. Activity of HRP using ABTS appears about four-fold higher than using pyrogallol. It is mainly used as a substrate in sensitive peroxidase assays.
2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS). Recommended for ELISA (microwell) procedures, not recommended for membrane applications.


  • Presentation: Light Green Powder
  • Extinction Coefficient (E1%): EmM(342 nM) > 37.4; EmM(225 nM) > 48.15; EmM(414 nM) = 36 (Lit.)
  • Soluble in water (5% w/v - clear, pale green solution).
  • Storage temperature: Store at +4 °C. Store Under Nitrogen. Protect from light.

Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010404
CAS Number: 30931-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4eff66a1e1fde06b7b705bbcc793a87b7fb0e96c&vid=4eff66a1e1fde06b7b705bbcc793a87b7fb0e96c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
UV/VIS: I max (water) 340 - 342, 224 - 228 nm
Moisture ≤5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR