N-Acetyl-L-cysteine ≥96%, white crystalline powder cell culture reagent

Supplier: 0000012207

0219460305 0219460325 0219460350
76177-018EA 42.35 USD
76177-018 76177-020 76177-022
N-Acetyl-L-cysteine ≥96%, white crystalline powder cell culture reagent
N-Acetyl-L-cysteine

N-Acetyl-L-cysteine is an acetylated amino acid with antioxidant and mucolytic properties for isolation of mycobacteria from sputum. As a mucolytic agent, N-Acetyl-L-cysteine serves to dissipate disulfide bonds across mucoproteins, loosening and clearing the viscosity of sputum.
N-Acetyl-L-cysteine acts as a free-radical scavenger and inhibits the production of reactive oxygen species which can damage cellular structures. It is critical for the production of Glutathione, a key antioxidant enzyme in the body. It has also been shown to induce apoptosis in vascular smooth muscle cells, indicating that these cells respond differently to changes in reduction-oxidation state than other tissues that are normally protected by the presence of antioxidants.
Antioxidant and mucolytic agent. Increases cellular pools of free radical scavengers. Reported to prevent apoptosis in neuronal cells but induce apoptosis in smooth muscle cells. Inhibits HIV replication. May serve as a substrate for microsomal glutathione transferase.


  • Presentation: White Crystalline Powder
  • pKa: 9.5 (Lit.)
  • Soluble in water: 100 mg/mL (with heating).

Formula: C₅H₉NO₃S
Melting Pt: 105…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004880
CAS Number: 616-91-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8927a5603825bed8b63c44a45269ca6e60eb79fe&vid=9dbb7bb8d9838324bd3f97e42d04eb3e6d4438fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥96%
FTIR Conforms to Standard
Optical Rotation +21.0° to +27.0° (USP soln)
Moisture ≤1.0%
pH (1%aq.) 2.0-2.8
Heavy Metals ≤0.001%
Residue on Ignition ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR