Tissue-Tek® Prisma® Plus Automated Slide Stainer, Special Stain Configuration, Sakura Finetek

Supplier: Sakura Finetek
6171
76176-848EA 83913.25 USD
76176-848
Tissue-Tek® Prisma® Plus Automated Slide Stainer, Special Stain Configuration, Sakura Finetek
Slide Stainers
High throughput slide stainer, capable of processing up to 530 slides per hour. Continuous loading of slides, with intuitive software to run multiple staining protocols simultaneously.

  • Capable of up to 50 user-defined staining protocols
  • Supplied with 2 addtional heating stations (30 to 70 °C) for special stains
  • Capable of prioritizing stat run over standard
  • Color touchscreen with choice of 50 colors for screen stain assignments
  • Barcode reader to track reagent usage and alerts for expired reagent
  • iSupport service link monitors service issues

Standard solution configuration means 30 reagent positions and 60 slides per run or expanded configuration is 52 positions and 20 slides per run.

Connect the Prisma® Plus with the Tissue-Tek® Film® or G2 coverslippers for a fully automated, integrated, walkaway staining process from slide baking to slide drying.

Certifications: IEC 61010-1 2nd ed., CAN/CSA C22.2 No. 61010-1, UL 61010-1.

Delivery information: Includes with operating manual, twenty-two 820 mL solution reservoirs, ten 20-slide baskets and handles, two heated solution reservoirs, ten 10-slide baskets and handles, eight 160 mL special stain reservoirs, four wash reservoirs, two fume filters, compact flash card and reagent lids.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR