Anti-P2X5 Polyclonal Antibody

Supplier: Boster Biological Technology
PB9305
76173-444EA 787.77 USD
76173-444
Anti-P2X5 Polyclonal Antibody
Antibodies
Polyclonal antibody for P2X5/P2RX5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. P2X5/P2RX5 information: Molecular Weight: 47205 MW; Subcellular Localization: Membrane; Multi-pass membrane protein; Tissue Specificity: Expressed at high levels in brain and immune system.

Type: Primary
Antigen: P2X5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR