Anti-KIT/SCFR Polyclonal Antibody

Supplier: Boster Biological Technology
PB9258
76173-350EA 787.77 USD
76173-350
Anti-KIT/SCFR Polyclonal Antibody
Antibodies
Polyclonal antibody for C KIT/KIT detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. C KIT/KIT information: Molecular Weight: 109865 MW; Subcellular Localization: Isoform 1: Cell membrane; Single-pass type I membrane protein; Tissue Specificity: Isoform 1 and isoform 2 are detected in spermatogonia and Leydig cells. Isoform 3 is detected in round spermatids, elongating spermatids and spermatozoa (at protein level). Widely expressed. Detected in the hematopoietic system, the gastrointestinal system, in melanocytes and in germ cells.

Type: Primary
Antigen: KIT/SCFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR