Anti-ACE/Cd143 Polyclonal Antibody

Supplier: Boster Biological Technology
PB9124
76172-944EA 793.53 USD
76172-944
Anti-ACE/Cd143 Polyclonal Antibody
Antibodies
Polyclonal antibody for CD143/ACE detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD143/ACE information: Molecular Weight: 149715 MW; Subcellular Localization: Angiotensin-converting enzyme, soluble form: Secreted; Tissue Specificity: Ubiquitously expressed, with highest levels in lung, kidney, heart, gastrointestinal system and prostate. Isoform Testis-specific is expressed in spermatocytes and adult testis.

Type: Primary
Antigen: ACE/Cd143
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR