Human TrkB PicoKine; ELISA Kit, Boster
Supplier: Boster Biological Technology
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human TrkB
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human Trkb/NTRK2 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates and cell lysates. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Trkb/NTRK2 tissue specificity: Isoform TrkB is expressed in the central and peripheral nervous system. In the central nervous system (CNS), expression is observed in the cerebral cortex, hippocampus, thalamus, choroid plexus, granular layer of the cerebellum, brain stem, and spinal cord. In the peripheral nervous system, it is expressed in many cranial ganglia, the ophthalmic nerve, the vestibular system, multiple facial structures, the submaxillary glands, and dorsal root ganglia. Isoform TrkB-T1 is mainly expressed in the brain but also detected in other tissues including pancreas, kidney and heart. Isoform TrkB-T-Shc is predominantly expressed in the brain. .
96wells/kit, with removable strips.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...