Anti-CHD2 IgG Polyclonal Antibody

Supplier: Boster Biological Technology
A04079
76171-804EA 945.34 USD
76171-804
Anti-CHD2 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Chromodomain-helicase-DNA-binding protein 2(CHD2) detection. Tested with WB, IHC-P in Human;Rat.

Type: Primary
Antigen: CHD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR