Anti-S100A7 IgG Polyclonal Antibody

Supplier: Boster Biological Technology
A02369-1
76171-610EA 787.77 USD
76171-610
Anti-S100A7 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Psoriasin detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: S100A7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR